Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Heatex™ Open 1 in. Core, Ink-Side-Out Printer Ribbon - R4300 Wax/Resin, White

Catalog No. 19104559 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-559 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-559 Supplier Brady™ Supplier No. 379828

This ink-side-out ribbon features an open 1 in. core for compatibility with a wide variety of thermal transfer printers.

  • One-color ribbon for printing a single color onto a label
  • R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals when printed on the appropriate Brady™ label material
  • Recommended for printing on paper and films that have a matte or rough surface, such as B-427 Self-Laminating Vinyl and B-499 Nylon Cloth from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance

Specifications

Product Type Printer Ribbon
Color White
For Use With (Equipment) BBP11 Label Printer, BBP12 Label Printer
Length (English) 984 ft.
Autoclavable Not Autoclavable
Description Printer Ribbon
Format Open-core Ribbon
Quantity 1 Roll
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.