Learn More
Heathrow Scientific™ Anti-Microbial Modular Glove Box Holder
GREENER_CHOICE
Catalog No. 22267076
Change view
Click to view available options
Color:
Blue
Catalog No. Color
22-267-076 Blue
1 options
This item is not returnable. View return policy
Catalog No. 22-267-076 Supplier Heathrow Scientific™ Supplier No. HS120258
  • $163.00
    $118.65 / Pack of 3
    Save $44.35
    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
This item is not returnable. View return policy
Infused with Microban™ for anti-microbial protection making this glove box holder ideal for use in clean rooms, hospitals, and microbiological work areas.
  • Infused with Microban™ for anti-microbial protection
  • Microban™ additive prevents growth of molds and bacteria
  • High-impact, durable resin is resistant to harsh cleaning solutions
  • Modular design allows single boxes, or horizontal stacking of multiple boxes
  • Compatible with most brands of disposable glove boxes
  • Mounting hardware is included
Specifications
Product Type Glove Box Holder
Color Blue
Dimensions (L x W x H) 10 x 6.1 x 3.8 in. (254 x 156 x 97 mm)
Holds Disposable Glove Boxes
Material Resin
Mounting Keyhole Slots
No. of Compartments 1
Product Style Front-loading with Opening
Description Modular Design
Capacity 1 Box
Includes Mounting hardware
Sterility Non-sterile