Learn More
ATCC Heat-inactivated; Monkeypox virus; Isolate: hMPXV/USA/MA001/2022
Supplier:  ATCC VR3371HK
Encompass
This product is a preparation of Monkeypox virus strain hMPX/USA/MA001/2022 (clade IIb) that has been inactivated by heating at 65°C for 30 minutes and is therefore unable to replicate. This heat-inactivated preparation is whole-genome sequenced and can be used as a process control in the development of Monkeypox molecular diagnostic tools and can also be used to make a dilution series to estimate the level of detection.
Catalog No. 50-238-3619