Learn More
ATCC Heat-inactivated SARS-CoV-2, Omicron variant strain USA/GA-EHC-2811C/2021 (ATCC VR-3347HK™)
Catalog No. 502383654
Encompass
Heat-inactivated SARS-CoV-2, Omicron variant strain USA/GA-EHC-2811C/2021 (ATCC VR-3347HK™) is a whole-genome sequenced, non-replicative preparation used as a process control in molecular assay development. Inactivated at 65°C for 30 minutes, it supports qPCR workflows and variant-specific detection.
Catalog No. 50-238-3654 Supplier ATCC Supplier No. VR3347HK
Heat-inactivated SARS-CoV-2, Omicron variant strain USA/GA-EHC-2811C/2021 (ATCC VR-3347HK™) is a whole-genome sequenced, non-replicative preparation used as a process control in molecular assay development. Inactivated at 65°C for 30 minutes, it supports qPCR workflows and variant-specific detection.