Learn More
ATCC Heat-inactivated; SARS-CoV-2; Omicron variant BA.5; Strain: USA/COR-22-063113/2022
Supplier:  ATCC VR3378HK
Encompass
This product is a preparation of Severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2) strain USA/COR-22-063113/2022 (Omicron variant) that has been inactivated by heating at 65°C for 30 minutes and is therefore unable to replicate. This heat-inactivated preparation is whole-genome sequenced and can be used as a process control in the development of SARS-CoV-2 molecular diagnostic tools and can also be used to make a dilution series to estimate the level of detection. Due to the dynamic nature of lineage determination using PANGO, it is recommended to confirm the provided GISAID information prior to initiation of work.
Catalog No. 50-238-1710