Learn More
Labtech Inc 5 liter water/oil Heating Bath
Catalog No. NC1471047
Encompass_Preferred
5 liter water/oil heating bath. Temperature range ambient to 180˚C, automatically turn off in dry bath condition
Catalog No. NC1471047 Supplier Labtech Inc Supplier No. HB03PLUS
5 liter water/oil heating bath. Temperature range ambient to 180˚C, automatically turn off in dry bath condition