Learn More
BUCHI Heating Bath B-305
SDP
Catalog No. 05405089 Shop All BUCHI Products
Change view
Click to view available options
Includes:
Heating Bath (B-305)
Heating Bath (B-305), Base (B-300)
Heating Bath (B-305), Base (B-300), Rotavapor R-21 Adapter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
05-405-089 Heating Bath (B-305)
05-405-090 Heating Bath (B-305), Base (B-300)
05-405-091 Heating Bath (B-305), Base (B-300), Rotavapor R-21 Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-089 Supplier BUCHI Supplier No. 11B305002
Add to Cart
Add to Cart
Compatible with evaporating flasks up to 5 liters.
  • Use with water or oil as heating media
  • For temperatures up to 220°C
Specifications
Material Stainless Steel
Type Heating Bath
For Use With (Application) Water or Oil
For Use With (Equipment) B-300, R-300
Includes Heating Bath (B-305)
Temperature (Metric) Operating ≤220°C
Volume (Metric) 5.5 L