Learn More
Chemglass Life Sciences Heating Blanket, 15 Liter Bio Reactor, Velcro Style with 2 prong Plug, 110 Volt, 383 Watt
SDP
Catalog No. 501294522
Encompass
Manufactured to fit snugly around Applikon-style, unjacketed vessels to supply uniform heat. Made of silicone rubber, all sizes are 110VAC with the wattage as listed. Velcro allows easy installation, even with the vessel in a support stand. Plugs are IEC standard 2-prong.Please Note: These Heaters Cannot Be Directly Plugged Into a Wall Receptacle and Require a Temperature Controller with a Thermocouple or RTD in Order to Properly Maintain and Reach a Controlled Temperature.
Catalog No. 50-129-4522 Supplier Chemglass Life Sciences Supplier No. CLS138512V
Manufactured to fit snugly around Applikon-style, unjacketed vessels to supply uniform heat. Made of silicone rubber, all sizes are 110VAC with the wattage as listed. Velcro allows easy installation, even with the vessel in a support stand. Plugs are IEC standard 2-prong.Please Note: These Heaters Cannot Be Directly Plugged Into a Wall Receptacle and Require a Temperature Controller with a Thermocouple or RTD in Order to Properly Maintain and Reach a Controlled Temperature.