Learn More
Chemglass Life Sciences Heating Blanket, 2 Liter Bio Reactor, Velcro Style with 2 prong Plug, 110 Volt, 110 Watt
SDP
Catalog No. 501897541
Encompass
Heating Blanket, 2 Liter Bio Reactor, Velcro Style with 2 prong Plug, 110 Volt, 110 Watt. Manufactured to fit snugly around Applikon-style, unjacketed vessels to supply uniform heat. Made of silicone rubber. Velcro allows easy installation, even with the vessel in a support stand. Plugs are IEC standard 2-prong.
Catalog No. 50-189-7541 Supplier Chemglass Life Sciences Supplier No. CLS137908V
Heating Blanket, 2 Liter Bio Reactor, Velcro Style with 2 prong Plug, 110 Volt, 110 Watt. Manufactured to fit snugly around Applikon-style, unjacketed vessels to supply uniform heat. Made of silicone rubber. Velcro allows easy installation, even with the vessel in a support stand. Plugs are IEC standard 2-prong.