Learn More
Andwin Scientific HEATING BLOCK 30 HOLE TAPERED
SDP
Catalog No. NC9623144
Encompass_Preferred
Modular Heating Block, 30-hole. Holds Tapered Microcentrifuge Tubes up to 0.5mL and 7.9mm dia. Solid aluminum, anodized blue modular block for producing maximum heat retention in and Signature Dry Block Heaters.
Catalog No. NC9623144 Supplier Andwin Scientific Supplier No. 13259000
Modular Heating Block, 30-hole. Holds Tapered Microcentrifuge Tubes up to 0.5mL and 7.9mm dia. Solid aluminum, anodized blue modular block for producing maximum heat retention in and Signature Dry Block Heaters.