Learn More
Research Products International Corp Heating Block, Holds 12x16mm Tubes
SDP
Catalog No. 501977726
Encompass
Heating Block Module for the Digital Dry Baths. Ideal block modules for use with Digital DryBath (sku 246201). Each block measures 3 1/2 x 3 x 2 1/2 inches high.
Catalog No. 50-197-7726 Supplier Research Products International Corp Supplier No. 246210
Heating Block Module for the Digital Dry Baths. Ideal block modules for use with Digital DryBath (sku 246201). Each block measures 3 1/2 x 3 x 2 1/2 inches high.