Learn More
Research Products International Corp Heating Block, Holds 20x13mm Tubes
SDP
Catalog No. 501977778
Encompass
Heating Block Module for the Digital Dry Baths. Ideal block modules for use with Digital DryBath (sku 246201). Each block measures 3 1/2 x 3 x 2 1/2 inches high.
Catalog No. 50-197-7778 Supplier Research Products International Corp Supplier No. 246208
Heating Block Module for the Digital Dry Baths. Ideal block modules for use with Digital DryBath (sku 246201). Each block measures 3 1/2 x 3 x 2 1/2 inches high.