Learn More
Research Products International Corp Heating Block Module, Holds 1 x 96 Well 0.2ml Microplate, Strips or Tubes
SDP
Catalog No. 50212158
Encompass
This heating block module is designed to handle plates, tube strips or 0.2ml tubes and are designed to fit both Digital and Analog Dry Baths.
Catalog No. 50-212-158 Supplier Research Products International Corp Supplier No. 110096
This heating block module is designed to handle plates, tube strips or 0.2ml tubes and are designed to fit both Digital and Analog Dry Baths.