Learn More
Research Products International Corp Heating Block Module for 13mm Test Tubes, 20 Tube Capacity
SDP
Catalog No. 50212151
Encompass
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.
Catalog No. 50-212-151 Supplier Research Products International Corp Supplier No. 110013
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.