Learn More
Research Products International Corp HEATING BLOCK, HOLDS 5 X 50 ML TUBES, 3 1/2 X 3 X 2 INCHES HIGH
SDP
Catalog No. 50550237
Encompass
Heating Block Module for the Digital Dry Baths. Ideal block modules for use with Digital DryBath (sku 246201). Each block measures 3 1/2 x 3 x 2 1/2 inches high.
Catalog No. 50-550-237 Supplier Research Products International Corp Supplier No. 246213
Heating Block Module for the Digital Dry Baths. Ideal block modules for use with Digital DryBath (sku 246201). Each block measures 3 1/2 x 3 x 2 1/2 inches high.