Learn More
Research Products International Corp Heating Block Module for Test Tubes, Combo Block holds 3 x 25mm Tubes, 5 x 13mm Tubes and 6 x 6mm Tubes
SDP
Catalog No. 50212154
Encompass
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.
Catalog No. 50-212-154 Supplier Research Products International Corp Supplier No. 110035
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.