Learn More
Julabo USA Inc CORIO CP-BC26 2 kW heater; +20…+200 °C operating range; ±0.02 °C stability; 8-27 L/min pump with 1.5-10.2 psi; 19-26 L tank; adjustable internal/external flow; USB & RS232 communication
SDP
Catalog No. 501904656
Encompass_Preferred
CORIO™ CP, The new generation of laboratory circulators from JULABO.CORIO™ CP heating circulators are suitable for temperature applications with circulation to an external apparatus or immersing a device in the bath tank. The CORIO™ CP circulator incorporates easy change-over for control of fluid circulation internally or externally. Unit fittings M16x1 male threaded adapters including a pair of 8 and 12 mm hose barbs. Systems include insulated stainless-steel tanks. Systems include cooling coil in bath tank and integrated drain port. Operating range from +20...+200 °C.Bright LED display with easy to use interface. USB and RS232 connections for external PC control and data-logging; PID temperature control provides ±0.02 °C stability. Adjustable centrifugal pump capacity of 8-27 L/min with pressure from 1.5-10.2 psi.Class III (FL) according to DIN 12876-1.
Catalog No. 50-190-4656 Supplier Julabo USA Inc Supplier No. 9013526.33
Add to Cart
Add to Cart
Request a Quote
CORIO™ CP, The new generation of laboratory circulators from JULABO.CORIO™ CP heating circulators are suitable for temperature applications with circulation to an external apparatus or immersing a device in the bath tank. The CORIO™ CP circulator incorporates easy change-over for control of fluid circulation internally or externally. Unit fittings M16x1 male threaded adapters including a pair of 8 and 12 mm hose barbs. Systems include insulated stainless-steel tanks. Systems include cooling coil in bath tank and integrated drain port. Operating range from +20...+200 °C.Bright LED display with easy to use interface. USB and RS232 connections for external PC control and data-logging; PID temperature control provides ±0.02 °C stability. Adjustable centrifugal pump capacity of 8-27 L/min with pressure from 1.5-10.2 psi.Class III (FL) according to DIN 12876-1.