Learn More
Description
Includes
PCR block, Polycarbonate lid
- Microprocessor control - The variable speed microprocessor control provides consistent, uniform shaking action. Microprocessor will display last set-point and will restart if power is interrupted.
- PID temperature controller - Maintains precise temperature control from 10°C below ambient to 65°C. Easy-to-use controls allow users to adjust temperature in 1°C increments.
- Triple eccentric drive - Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.
- LED display - Touch pad controls with easy-to-read, independent LED displays for temperature, speed and time allow operator to view all settings at once. Provides repeatable and accurate results every time and is easily visible across lab benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.
- Temperature calibration mode - Allows user to calibrate unit to an external temperature device
- Applications - ELISA, Enzyme Activity Analysis, Enzyme cutting, DNA ligation, PCR
Specifications
Specifications
| Orbit | 3 mm |
| Speed Range | 150 to 1200 rpm (PCR plate) |
| Electrical Requirements | 230 V,1 A, 450 W, Plug Type: EU, UK, CN |
| Voltage | 230 V |
| Temperature Range (Metric) | Ambient -10° to 95°C (Lowest Possible Temp: 5°C) |
| Certifications/Compliance | CE |
| Plug Type | EU, UK, CN Plug |
| Material | Aluminum Platform |
| Dimensions (D x W x H) Interior | 278 x 222 x 155 mm |
| Dimensions (LxW) Platform | 7 x 5.5 in. (180 x 140 mm) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.