Learn More
Chemglass Life Sciences Chemglass Glas-Col Series TM 1000 mL Spherical Heating Mantle, 230 V Euro Plug (CG-12000-08CE)
SDP
Catalog No. NC3786441
Encompass_Preferred
The Chemglass Glas-Col Series TM 1000 mL spherical heating mantle (CG-12000-08CE) is a hemispherical, aluminum-housed mantle designed to heat 1 L spherical flasks. It provides 380 W of heating power and is supplied with a 230 V Euro plug; the unit is intended for use with an external temperature controller (thermocouple or RTD) and is not designed to be directly connected to a wall receptacle.
  • 1000 mL capacity.
  • 380 W heating power.
  • 230 V Euro plug.
  • Hemispherical aluminum housing.
  • Requires external temperature controller with thermocouple or RTD.
  • Covers bottom half of spherical flask to allow visibility of contents.
  • Not intended to be directly plugged into a wall receptacle.
Catalog No. NC3786441 Supplier Chemglass Life Sciences Supplier No. CG1200008CE
The Chemglass Glas-Col Series TM 1000 mL spherical heating mantle (CG-12000-08CE) is a hemispherical, aluminum-housed mantle designed to heat 1 L spherical flasks. It provides 380 W of heating power and is supplied with a 230 V Euro plug; the unit is intended for use with an external temperature controller (thermocouple or RTD) and is not designed to be directly connected to a wall receptacle.
  • 1000 mL capacity.
  • 380 W heating power.
  • 230 V Euro plug.
  • Hemispherical aluminum housing.
  • Requires external temperature controller with thermocouple or RTD.
  • Covers bottom half of spherical flask to allow visibility of contents.
  • Not intended to be directly plugged into a wall receptacle.