Learn More
Chemglass Life Sciences 1000mL Glas-Col Heating Mantle, Series TM, Spherical, 380 Watts
SDP
Catalog No. 50401777
Encompass_Preferred
Mantle, Flask; Chemglass; Glas-Col; Series TM; Spherical; Covers only bottom half of flask; Larger sizes have multiple circuits for ease of temp. ctrl when flask is less than half full; Cap.: 1000mL; 115V, 380W
Catalog No. 50-401-777 Supplier Chemglass Life Sciences Supplier No. CG1200008
Mantle, Flask; Chemglass; Glas-Col; Series TM; Spherical; Covers only bottom half of flask; Larger sizes have multiple circuits for ease of temp. ctrl when flask is less than half full; Cap.: 1000mL; 115V, 380W