Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0670976
Encompass_Preferred
Fabric heating mantle for 100ml spherical flask, to 400 degrees C maximum internal operating temperature. 80W, 115V, CSA
Catalog No. NC0670976 Supplier Glas Col LLC Supplier No. 100A O396
Fabric heating mantle for 100ml spherical flask, to 400 degrees C maximum internal operating temperature. 80W, 115V, CSA