Learn More
Chemglass Life Sciences Reactor Capacity: 10 Liter, Watts: 450
SDP
Catalog No. CG1201210
Encompass_Preferred
Heating Mantle, Lower Portion, Unjacketed Cylindrical Vessel, Reactor Capacity: 10 Liter, Watts: 450. For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels. TM Series aluminum housed glass fabric heating mantle with hole centered in the bottom. Mantle is 115 volts and is supplied complete with a separable 4ft 2-wire cord and locking connector.
Catalog No. CG1201210 Supplier Chemglass Life Sciences Supplier No. CG1201210
Heating Mantle, Lower Portion, Unjacketed Cylindrical Vessel, Reactor Capacity: 10 Liter, Watts: 450. For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels. TM Series aluminum housed glass fabric heating mantle with hole centered in the bottom. Mantle is 115 volts and is supplied complete with a separable 4ft 2-wire cord and locking connector.