Learn More
Chemglass Life Sciences Reactor Capacity: 10 Liter, Watts: 720
SDP
Catalog No. CG1001310
Encompass_Preferred
For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels. Glass fabric heating mantle with a lengthwise slit held together with a hook and lace closure. Mantle is 115 volts is supplied complete with a separable 4ft 2-wire cord and locking connector. Heating Mantle, Upper Portion, Unjacketed Cylindrical Vessel, Reactor Capacity: 10 Liter, Watts: 720.
Catalog No. CG1001310 Supplier Chemglass Life Sciences Supplier No. CG1001310
For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels. Glass fabric heating mantle with a lengthwise slit held together with a hook and lace closure. Mantle is 115 volts is supplied complete with a separable 4ft 2-wire cord and locking connector. Heating Mantle, Upper Portion, Unjacketed Cylindrical Vessel, Reactor Capacity: 10 Liter, Watts: 720.