Learn More
Azzota Corp Heating Mantle, Capacity: 2000Ml, Heating Power: 350W, Maximum Temp. 380C (716F)
Catalog No. 501612853
Encompass_Preferred
Heating Mantle, Capacity: 2000ml, Heating Power: 350W, Maximum Temp. 380C (716F)
Catalog No. 50-161-2853 Supplier Azzota Corp Supplier No. HM2000
Heating Mantle, Capacity: 2000ml, Heating Power: 350W, Maximum Temp. 380C (716F)