Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC9980376
Encompass_Preferred
Fabric heating mantle for 2L spherical flask to 400 degrees C maximum internal operating temperature. 500W, 115V, CSA
Catalog No. NC9980376 Supplier Glas Col LLC Supplier No. 100A O410
Fabric heating mantle for 2L spherical flask to 400 degrees C maximum internal operating temperature. 500W, 115V, CSA