Learn More
Azzota Corp Heating mantle 250ml 150W 220V
Catalog No. 5000887231
Encompass_Preferred
Heating Mantle Capacity 250ml Heating Power 150W Maximum Temp 380C (716F) 220V
Catalog No. 50-008-87231 Supplier Azzota Corp Supplier No. HM250220V
Heating Mantle Capacity 250ml Heating Power 150W Maximum Temp 380C (716F) 220V