Learn More
Azzota Corp Electric Heating Mantle 250ml
Catalog No. 501179481
Encompass_Preferred
Electric Heating Mantle, Capacity: 250ml, Maximum temperature 420C (790F); Dimension (mm): Dia 220mm x 165mm; Weight: 6lbs (2.5Kg)
Catalog No. 50-117-9481 Supplier Azzota Corp Supplier No. HM250
Electric Heating Mantle, Capacity: 250ml, Maximum temperature 420C (790F); Dimension (mm): Dia 220mm x 165mm; Weight: 6lbs (2.5Kg)