Learn More
Chemglass Life Sciences Reactor Capacity: 30 Liter, Watts: 1250
SDP
Catalog No. CG1001330
Encompass_Preferred
For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels. Glass fabric heating mantle with a lengthwise slit held together with a hook and lace closure. Mantle is 115 volts is supplied complete with a separable 4ft 2-wire cord and locking connector. Heating Mantle, Upper Portion, Unjacketed Cylindrical Vessel, Reactor Capacity: 30 Liter, Watts: 1250.
Catalog No. CG1001330 Supplier Chemglass Life Sciences Supplier No. CG1001330
For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels. Glass fabric heating mantle with a lengthwise slit held together with a hook and lace closure. Mantle is 115 volts is supplied complete with a separable 4ft 2-wire cord and locking connector. Heating Mantle, Upper Portion, Unjacketed Cylindrical Vessel, Reactor Capacity: 30 Liter, Watts: 1250.