Learn More
Ace Glass, Inc. Heating mantle, aluminum, 3L, 4.5x11in, CSA rated
SDP
Catalog No. 50977416
Encompass
3L aluminum-housed, table model mantle provides durability and strength to support the weight of the vessel. Designed specifically for cylindrical reaction kettles. Maximum internal operating temperature is 450 C. Dimensions: 4.5x 11. CSA rated.
Catalog No. 50-977-416 Supplier Ace Glass, Inc. Supplier No. 1207512
3L aluminum-housed, table model mantle provides durability and strength to support the weight of the vessel. Designed specifically for cylindrical reaction kettles. Maximum internal operating temperature is 450 C. Dimensions: 4.5x 11. CSA rated.