Learn More
Azzota Corp Heating mantle 500ml 250W 220V
Catalog No. 5000887256
Encompass_Preferred
Heating Mantle Capacity 500ml Heating Power 250W Maximum Temp 380C 220V
Catalog No. 50-008-87256 Supplier Azzota Corp Supplier No. HM500220V
Heating Mantle Capacity 500ml Heating Power 250W Maximum Temp 380C 220V