Learn More
Cole-Parmer Glas-Col 100A O406 Flexible Fabric Heating Mantle, 500 ml, 270 Watts, 120 V
Catalog No. NC9823123 Shop All Cole-Parmer Products
Encompass_Preferred
Hemispherical heating mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel extension supports (sold separately) are required to support mantles; do not place mantle directly on ring stand. Mantles require a controlled electrical power source to prevent overheating.
Catalog No. NC9823123 Supplier Cole-Parmer Supplier No. 3622530
Hemispherical heating mantles feature insulated glass fabric that conforms to unusual vessel shapes. Steel extension supports (sold separately) are required to support mantles; do not place mantle directly on ring stand. Mantles require a controlled electrical power source to prevent overheating.