Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC0130022
Encompass_Preferred
Fabric hemispherical mantle for 500ml spherical flask, heats to 400 degrees C maximum internal operating temperatue, 270W, 115V CSA
Catalog No. NC0130022 Supplier Glas Col LLC Supplier No. 100A O406
Fabric hemispherical mantle for 500ml spherical flask, heats to 400 degrees C maximum internal operating temperatue, 270W, 115V CSA