Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC9920334
Encompass_Preferred
Aluminum housed heating mantle for 22L spherical flask to 400 degrees C maximum internal operating temperature, 2 circuits @ 770W, 115V, CSAThe Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. mantles are available with a bottom hole for flask drains.
Catalog No. NC9920334 Supplier Glas Col LLC Supplier No. 100B TM118
Aluminum housed heating mantle for 22L spherical flask to 400 degrees C maximum internal operating temperature, 2 circuits @ 770W, 115V, CSAThe Series TM mantle covers only the flask's bottom half, letting you see its full contents. Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full. mantles are available with a bottom hole for flask drains.