Learn More
Chemglass Life Sciences 100mL Glas-Col Heating Mantle, Series O, Hemispherical, 80 Watts
SDP
Catalog No. CG1000002
Encompass_Preferred
100mL Glas-Col Heating Mantle, Series O, Hemispherical, 80 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.
Catalog No. CG1000002 Supplier Chemglass Life Sciences Supplier No. CG1000002
100mL Glas-Col Heating Mantle, Series O, Hemispherical, 80 Watts. The Glas-Col Series O mantle is a hemispherical style, meaning it covers only the bottom half of the flask. This allows full view of contents. The mantle accommodates single and multi-flasks. It should be used with the Series O supports. All mantles are 115 Volts and are supplied with a separable 4 foot, 2 wire cord and locking connector. For Series O supports see CG-10010 or CG-10012.