Learn More
Cole-Parmer Electrothermal Digi-Mantle Stirring/Heating Mantle, 500 mL; 115 V
Catalog No. 501960385 Shop All Cole-Parmer Products
Encompass
Designed to provide a comprehensive answer to heating and stirring fluids in various vessel formats. This product is modular in design, allowing for complete interchangeability between mantles of various sizes. When the temperature probe is fitted, the heating is controlled via the temperature feedback circuit and the actual temperature is displayed on the controller. A temperature probe may be used to gauge the temperature of the flask contents more accurately (up to 220°C) Choose from three heater cartridge sizes, accommodating 100 to 250 mL, 500 mL, or 1000 mL round bottom flasks.
Catalog No. 50-196-0385 Supplier Cole-Parmer Supplier No. 3620421
Designed to provide a comprehensive answer to heating and stirring fluids in various vessel formats. This product is modular in design, allowing for complete interchangeability between mantles of various sizes. When the temperature probe is fitted, the heating is controlled via the temperature feedback circuit and the actual temperature is displayed on the controller. A temperature probe may be used to gauge the temperature of the flask contents more accurately (up to 220°C) Choose from three heater cartridge sizes, accommodating 100 to 250 mL, 500 mL, or 1000 mL round bottom flasks.