Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1020160
Encompass_Preferred
Fabric heating mantle for 250ml Griffin beaker to 400 degrees C maximum internal operating temperature. 140W, 115V, CSA.
Catalog No. NC1020160 Supplier Glas Col LLC Supplier No. 100A O606
Fabric heating mantle for 250ml Griffin beaker to 400 degrees C maximum internal operating temperature. 140W, 115V, CSA.