Learn More
Braintree Scientific Heating Plate 1.5""""W X ?""""H X 4""""L
SDP
Catalog No. 501954939
Encompass
Heating Plate, HP-1M Direct current heating plate for small rodents(mice and rats). Standard plate is black anodized aluminum and measures 4 3/8W" x 8.5" with small feet giving it an elevation of 1/4" above the surface it sits on, such as a table or stereotaxic stage. It is easily cleaned with alcohol. It has a feedback sensor to prevent overheating when used with our TCAT-2 series of controllers. Custom sizes are also available with longer lead times, call for quote. Weight: 0.5 lbs. Power: 25 watts Temperature Feedback Sensor: 100 Ohm RTD **Sold for research purposes only and not for human use**
Catalog No. 50-195-4939 Supplier Braintree Scientific Supplier No. HP4M
Heating Plate, HP-1M Direct current heating plate for small rodents(mice and rats). Standard plate is black anodized aluminum and measures 4 3/8W" x 8.5" with small feet giving it an elevation of 1/4" above the surface it sits on, such as a table or stereotaxic stage. It is easily cleaned with alcohol. It has a feedback sensor to prevent overheating when used with our TCAT-2 series of controllers. Custom sizes are also available with longer lead times, call for quote. Weight: 0.5 lbs. Power: 25 watts Temperature Feedback Sensor: 100 Ohm RTD **Sold for research purposes only and not for human use**