Learn More
World Precision Instrument Heating Plates for Animal Temperature Controller, 15x4 cm
SDP
Catalog No. 502521237
Encompass
The Heating plates come in 3 sizes: 15x4cm, 15x10cm and 15x25cm and are for use with the Animal Temperature Controller (ATC2000).
Catalog No. 50-252-1237 Supplier World Precision Instrument Supplier No. 61840
The Heating plates come in 3 sizes: 15x4cm, 15x10cm and 15x25cm and are for use with the Animal Temperature Controller (ATC2000).