Learn More
World Precision Instrument Heating Plates for Animal Temperature Controller, 15x10 cm
SDP
Catalog No. 502521238
Encompass
The Heating plates come in 3 sizes: 15x4cm, 15x10cm and 15x25cm and are for use with the Animal Temperature Controller (ATC2000).
Catalog No. 50-252-1238 Supplier World Precision Instrument Supplier No. 61830
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Heating plates come in 3 sizes: 15x4cm, 15x10cm and 15x25cm and are for use with the Animal Temperature Controller (ATC2000).