Learn More
Grainger BriskHeat BS0 Silicone Rubber Heating Tape, 0.5 in., 12 ft., 313 W, 1 ea.
Catalog No. 502769635
Encompass_Preferred
These silicone rubber heating tapes provide an ideal, flexible solution for rapid, direct heating of flat or curved surfaces. Designed for use on apparatuses, gas tubing, and valves, they feature a moisture and chemical-resistant silicone rubber construction for durability in demanding environments.
  • Maximum exposure temperature of 232°C (450°F)
  • Fiberglass knitted and braided construction for increased flexibility
  • Multi-stranded resistance wire ensures long-term durability
  • Power leads located on the same end for simplified installation
  • Ingress protection rating of IP66
  • Compatible with both conductive and non-conductive surfaces
Catalog No. 50-276-9635 Supplier Grainger Supplier No. 13P794
These silicone rubber heating tapes provide an ideal, flexible solution for rapid, direct heating of flat or curved surfaces. Designed for use on apparatuses, gas tubing, and valves, they feature a moisture and chemical-resistant silicone rubber construction for durability in demanding environments.
  • Maximum exposure temperature of 232°C (450°F)
  • Fiberglass knitted and braided construction for increased flexibility
  • Multi-stranded resistance wire ensures long-term durability
  • Power leads located on the same end for simplified installation
  • Ingress protection rating of IP66
  • Compatible with both conductive and non-conductive surfaces