Learn More
Grainger BriskHeat BS0 Silicone Heating Tape, Silicone Rubber, 1 in. x 12 ft., 120V
Catalog No. 502769639
Encompass_Preferred
These silicone rubber heating tapes feature a flexible, moisture-resistant design ideal for providing fast, efficient direct heating to flat or curved surfaces. Constructed with a silicone rubber outer sheath and a fiberglass knitted and braided inner structure, they are suitable for use on metal apparatuses, gas tubing, and valves requiring temperatures up to 232°C (450°F).
  • Chemical, moisture, and radiation resistant
  • Non-conductive silicone rubber construction
  • IP66 rated for environmental protection
  • Requires an external controller
  • Ungrounded electrical configuration
Catalog No. 50-276-9639 Supplier Grainger Supplier No. 13P801
These silicone rubber heating tapes feature a flexible, moisture-resistant design ideal for providing fast, efficient direct heating to flat or curved surfaces. Constructed with a silicone rubber outer sheath and a fiberglass knitted and braided inner structure, they are suitable for use on metal apparatuses, gas tubing, and valves requiring temperatures up to 232°C (450°F).
  • Chemical, moisture, and radiation resistant
  • Non-conductive silicone rubber construction
  • IP66 rated for environmental protection
  • Requires an external controller
  • Ungrounded electrical configuration