Learn More
Grainger BriskHeat Silicone Heating Tape, 120V, 940W, 1 in x 18 ft
Catalog No. 502769342
Encompass_Preferred
The XtremeFLEX® BS0 silicone rubber heating tape provides a versatile, flexible solution for rapid and efficient direct heating on both flat and curved surfaces. Designed for demanding environments, this heating tape is engineered for use with metal apparatuses, gas tubing, and valves requiring consistent temperature management up to 450°F (232°C).
  • Chemical, moisture, and radiation resistant construction
  • Flexible, fiberglass-reinforced silicone rubber design
  • Includes 24 in. (61 cm) power leads for convenient installation
  • Non-conductive surface ideal for various industrial applications
  • Requires external temperature controller for operation
  • Meets CE, REACH, and RoHS standards
Catalog No. 50-276-9342 Supplier Grainger Supplier No. 13P804
The XtremeFLEX® BS0 silicone rubber heating tape provides a versatile, flexible solution for rapid and efficient direct heating on both flat and curved surfaces. Designed for demanding environments, this heating tape is engineered for use with metal apparatuses, gas tubing, and valves requiring consistent temperature management up to 450°F (232°C).
  • Chemical, moisture, and radiation resistant construction
  • Flexible, fiberglass-reinforced silicone rubber design
  • Includes 24 in. (61 cm) power leads for convenient installation
  • Non-conductive surface ideal for various industrial applications
  • Requires external temperature controller for operation
  • Meets CE, REACH, and RoHS standards