Learn More
Grainger Briskheat Heating Tape, Silicone Rubber, 2 in x 10 ft, 120V, 1 ea
Catalog No. 502773056
Encompass_Preferred
These silicone rubber heating tapes with adjustable thermostat control are designed for process temperature control, freeze protection, and supplemental heating applications. The flexible tape wraps easily around the desired object to maintain a set temperature ranging from 10°C to 218°C (50°F to 425°F).
  • Adjustable thermostat control from 10°C to 218°C (50°F to 425°F)
  • Moisture and chemical resistant construction
  • Suitable for use on both glass and metal surfaces
  • Includes 120V power cord with NEMA 1-15 plug
  • Maximum exposure temperature of 232°C (450°F)
  • IP54 rated for protection
  • Reusable design
Catalog No. 50-277-3056 Supplier Grainger Supplier No. 13P862
These silicone rubber heating tapes with adjustable thermostat control are designed for process temperature control, freeze protection, and supplemental heating applications. The flexible tape wraps easily around the desired object to maintain a set temperature ranging from 10°C to 218°C (50°F to 425°F).
  • Adjustable thermostat control from 10°C to 218°C (50°F to 425°F)
  • Moisture and chemical resistant construction
  • Suitable for use on both glass and metal surfaces
  • Includes 120V power cord with NEMA 1-15 plug
  • Maximum exposure temperature of 232°C (450°F)
  • IP54 rated for protection
  • Reusable design