Learn More
Grainger BriskHeat BS0 Heating Tape, Silicone Rubber, 120V, 2 in x 12 ft
Catalog No. 502769573
Encompass_Preferred
These silicone rubber heating tapes provide a flexible and efficient solution for surface heating on both flat and curved surfaces. Designed for rapid heat transfer, these tapes are suitable for use on metal surfaces, apparatuses, gas tubing, and valves, offering reliable performance up to 232°C.
  • Chemical, moisture, and radiation resistant
  • Non-conductive silicone rubber construction
  • Maximum exposure temperature of 232°C (450°F)
  • Power density of 4.3 watts/in2
  • IP66 rated for durability in demanding environments
  • Compatible with a wide range of standard heating controllers
Catalog No. 50-276-9573 Supplier Grainger Supplier No. 13P811
These silicone rubber heating tapes provide a flexible and efficient solution for surface heating on both flat and curved surfaces. Designed for rapid heat transfer, these tapes are suitable for use on metal surfaces, apparatuses, gas tubing, and valves, offering reliable performance up to 232°C.
  • Chemical, moisture, and radiation resistant
  • Non-conductive silicone rubber construction
  • Maximum exposure temperature of 232°C (450°F)
  • Power density of 4.3 watts/in2
  • IP66 rated for durability in demanding environments
  • Compatible with a wide range of standard heating controllers