Learn More
Grainger BriskHeat Heating Tape, Silicone Rubber, 120 V, 2 in. Width, 18 ft. Length, 1 ea
Catalog No. 502772952
Encompass_Preferred
This XtremeFLEX® BS0 series silicone rubber heating tape provides a flexible and efficient heating solution for flat or curved surfaces. Engineered for versatile industrial applications, it is suitable for use on apparatuses, gas tubing, and valves requiring rapid and reliable direct heat.
  • Maximum exposure temperature of 450°F (232°C)
  • Durable silicone rubber extruded outer sheath
  • Fiberglass knitted and braided inner construction
  • Chemical, moisture, and radiation resistant
  • Non-conductive design with integrated grounded wire
  • IP66 rating for environmental protection
  • Compatible with metal surfaces
  • Certifications include CE, REACH, and RoHS
Catalog No. 50-277-2952 Supplier Grainger Supplier No. 13P814
This XtremeFLEX® BS0 series silicone rubber heating tape provides a flexible and efficient heating solution for flat or curved surfaces. Engineered for versatile industrial applications, it is suitable for use on apparatuses, gas tubing, and valves requiring rapid and reliable direct heat.
  • Maximum exposure temperature of 450°F (232°C)
  • Durable silicone rubber extruded outer sheath
  • Fiberglass knitted and braided inner construction
  • Chemical, moisture, and radiation resistant
  • Non-conductive design with integrated grounded wire
  • IP66 rating for environmental protection
  • Compatible with metal surfaces
  • Certifications include CE, REACH, and RoHS