Learn More
Grainger BriskHeat HSTAT Heating Tape, Silicone Rubber, 218°C, 24 in.
Catalog No. 502769345
Encompass_Preferred
The HSTAT series heating tape is a portable, silicone rubber heating solution designed for diverse applications including process temperature maintenance, freeze protection, and de-icing of pipes and valves. This all-in-one, plug-and-play system features an integrated adjustable thermostat for precise thermal management in laboratory and industrial environments.
  • Adjustable thermostat control from 50°F to 425°F (10°C to 218°C)
  • Includes 1.83 m (6 ft) power cord with NEMA 1-15P 2-prong plug
  • Moisture and chemical resistant silicone rubber construction
  • Suitable for use on glass and metal surfaces
  • IP54 rated for protection against ingress
  • Portable and reusable design
Catalog No. 50-276-9345 Supplier Grainger Supplier No. 13P858
The HSTAT series heating tape is a portable, silicone rubber heating solution designed for diverse applications including process temperature maintenance, freeze protection, and de-icing of pipes and valves. This all-in-one, plug-and-play system features an integrated adjustable thermostat for precise thermal management in laboratory and industrial environments.
  • Adjustable thermostat control from 50°F to 425°F (10°C to 218°C)
  • Includes 1.83 m (6 ft) power cord with NEMA 1-15P 2-prong plug
  • Moisture and chemical resistant silicone rubber construction
  • Suitable for use on glass and metal surfaces
  • IP54 rated for protection against ingress
  • Portable and reusable design