Learn More
Grainger BRISKHEAT Heating Tape, 10 ft, 3x24"
Supplier:  Grainger 13P863
Encompass_Preferred
The BriskHeat Heating Tape is a 10 foot long self-regulating heating tape that provides freeze protection for pipes and valves. It has a maximum temperature of 218°F and can be cut to length. The heating tape features an integrated ground plug and requires a 120V power source. It has a built-in thermostat that adjusts heat output based on ambient temperature. The heating tape is made of fluoropolymer and stainless steel and can be used for indoor or outdoor applications. It is UL and CSA approved. The heating tape measures 0.2" W x 0.08" H and weighs 1 lb.
Catalog No. NC2621505