Learn More
Grainger BriskHeat Heating Tape, BIH Series, Grounded, 1 in. x 96 in., 1 ea.
Catalog No. 502769574
Encompass_Preferred
These grounded, heavy-insulated heating tapes provide reliable, high-temperature heat for a variety of laboratory and industrial applications. Designed with a patented grounded heating element, they are suitable for use on electrically conductive surfaces such as metal tubing or valves. The tapes are constructed from polyimide film wrapped in a fiberglass knitted and braided sheath to offer both durability and excellent flexibility.
  • Patented grounded heating element meets NEC 427.23 requirements
  • Maximum exposure temperature of 250°C (482°F)
  • Power density of 9.6 W/in2
  • Flexible fiberglass construction for easy installation
  • Requires a temperature controller (sold separately) for safe operation
Catalog No. 50-276-9574 Supplier Grainger Supplier No. 13P778
These grounded, heavy-insulated heating tapes provide reliable, high-temperature heat for a variety of laboratory and industrial applications. Designed with a patented grounded heating element, they are suitable for use on electrically conductive surfaces such as metal tubing or valves. The tapes are constructed from polyimide film wrapped in a fiberglass knitted and braided sheath to offer both durability and excellent flexibility.
  • Patented grounded heating element meets NEC 427.23 requirements
  • Maximum exposure temperature of 250°C (482°F)
  • Power density of 9.6 W/in2
  • Flexible fiberglass construction for easy installation
  • Requires a temperature controller (sold separately) for safe operation