Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Heating Transfer Block
GREENER_CHOICESDP

Catalog No. 31502132 Shop All Kemtech Products
Change view
Click to view available options
Capacity (Metric):
3 mL, 10 mL and 25 mL
3, 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
31-502-132 3, 5 mL
31-502-133 3 mL, 10 mL and 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-132 Supplier Synthware™ Supplier No. H200001
Add to Cart
Add to Cart

Used in place of glass sand in microscale experiments

Specifications

Color Silver
Capacity (Metric) 3, 5 mL
Material Aluminum
For Use With (Application) Organic Synthesis
For Use With (Equipment) Holds 3 and 5 mL heavywall reaction vials, 4 Craig crystallization tubes, 1 standard thermometer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.