Learn More
Integra Biosciences Corp HEATMAG MODULE
SDP
Catalog No. NC3373197
Encompass
  • Optimized for magnetic bead purification workflows
  • Temperature range from ambient to 65 °C
  • Exceptional magnet strength for effective separation
  • Automated vertical magnet movement
  • Adjustable magnet height for various applications
  • Integration options: standalone, semi-automated, fully automated
  • Compatible with ASSIST PLUS, VIAFLO 96, MINI 96, VIAFLO 384 pipetting platforms
  • Accepts 1.5 ml tubes (4 x 6) and 96 well PCR plates
  • Key applications include lysis, elution, nucleic acid extraction
  • Streamlined molecular biology and proteomics protocols
  • Fast and precise bead collection
  • Minimal bead carryover
  • High yield for purification processes
  • Full walk-away automation with ASSIST PLUS
  • One-button control for easy operation
Catalog No. NC3373197 Supplier Integra Biosciences Corp Supplier No. 4901
  • Optimized for magnetic bead purification workflows
  • Temperature range from ambient to 65 °C
  • Exceptional magnet strength for effective separation
  • Automated vertical magnet movement
  • Adjustable magnet height for various applications
  • Integration options: standalone, semi-automated, fully automated
  • Compatible with ASSIST PLUS, VIAFLO 96, MINI 96, VIAFLO 384 pipetting platforms
  • Accepts 1.5 ml tubes (4 x 6) and 96 well PCR plates
  • Key applications include lysis, elution, nucleic acid extraction
  • Streamlined molecular biology and proteomics protocols
  • Fast and precise bead collection
  • Minimal bead carryover
  • High yield for purification processes
  • Full walk-away automation with ASSIST PLUS
  • One-button control for easy operation